Injection needles
100 mm, 0,60 mm
Catalog # 612-0401
Supplier: ROMETSCH
Jump to:
Product Details & Documents
Rust-proof, reusable injection needles with nickel-plated Luer lock fitting.
- Cannulas for puncturing and injecting
- Normal bevelled tip
Specifications
- Ø:0,60 mm
- Lunghezza:100 mm