Orders LinkContactUsLinkComponent
Injection needles
Injection needles
100 mm, 0,60 mm
Catalog # 612-0401
Supplier:   ROMETSCH
Injection needles
Catalog # 612-0401
Supplier:  ROMETSCH
Injection needles
Injection needles
100 mm, 0,60 mm
Price
SFr. 56.70
Log in for availability
1 * 12 ST
Product Details & Documents
Rust-proof, reusable injection needles with nickel-plated Luer lock fitting.
  • Cannulas for puncturing and injecting
  • Normal bevelled tip
Specifications
  • Ø:
    0,60 mm
  • Lunghezza:
    100 mm