Injection needles
200 mm, 1,0 mm
Codice catalogo 612-0365
Nome del fornitore: ROMETSCH
Vai a
Product Details & Documents
Rust-proof, reusable injection needles with nickel-plated Luer lock fitting.
- Cannulas for puncturing and injecting
- Normal bevelled tip
Specifiche
- Ø:1,0 mm
- Lunghezza:200 mm