Orders LinkContactUsLinkComponent
Injection needles
Injection needles
150 mm, 2,0 mm
Codice catalogo 612-0360
Nome del fornitore:   ROMETSCH
Injection needles
Codice catalogo 612-0360
Nome del fornitore:  ROMETSCH
Injection needles
Injection needles
150 mm, 2,0 mm
Price
SFr. 153.00
Log in per disponibilità
1 * 12 ST
Product Details & Documents
Rust-proof, reusable injection needles with nickel-plated Luer lock fitting.
  • Cannulas for puncturing and injecting
  • Normal bevelled tip
Specifiche
  • Ø:
    2,0 mm
  • Lunghezza:
    150 mm