Diagonal cutters
Diagonal cutter, chrome plated, handles insulated with multi-component grips
Catalog # 238-5310
Supplier: KNIPEX WERKE
Jump to:
Product Details & Documents
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
- High quality material and precise workmanship for long service life
- Precision cutting edges for soft and hard wire
- Cutting edges additionally induction hardened
Certifications: DIN ISO 5749. VDE-tested.
Specifications
- Descrizione:Diagonal cutter, chrome plated, handles insulated with multi-component grips
- Seq:02