Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Scalpels, ceramic

10, 130 mm, 20 mm
Codice catalogo 233-5091
Nome del fornitore:   SLG Kunststoff
Scalpels, ceramic
Codice catalogo 233-5091
Nome del fornitore:  SLG Kunststoff
Scalpels, ceramic
Scalpels, ceramic

10, 130 mm, 20 mm

Price
SFr. 253.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Particularly high quality and durable hard ceramic material for long lasting, excellent sharpness. No metal parts, thus also suitable for trace analysis. Super-lightweight, sturdy, ergonomic handle for safe and easy handling.

  • Anti-magnetic, anti-static, corrosion-free, dishwasher safe, chemically resistant to aggressive media
  • Extremely heat resistant, low thermal conductivity
  • Regrinding is not necessary
Specifiche
  • Tipo:
    10
  • Lunghezza:
    130 mm
  • Seq:
    02
  • Lama:
    20 mm