Connectors, PP
Connector, straight, male luer lock/tubing nipple, 4,0 mm
Codice catalogo 229-0518
Nome del fornitore: PIEPER FILTER
Vai a
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Documents
Specifiche
- Ø int.tubo:4,0 mm
- Descrizione:Connector, straight, male luer lock/tubing nipple
- Seq:04