Thermosafe Cool Boxes
Roller for cooling boxes, -
Katalognr. 216-7002
Anbieter: HONIGBERG
Direkt zu
Product Details & Documents
PE, grey.
- Insulated with 5,75 cm thick plastic foam
- Hinged lid with robust hinges and metal catch, one sturdy carrying handle with the 35 litre model and two with the 75 litre model
- Aluminium roller base, with 4 moving rollers, available separately
Large capacity cool boxes for the long-term temperature control of temperature-sensitive materials and samples. Cooled with cooling pouches 216-7010 to 216-7013 or dry ice.
Technische Daten
- Capacità:-
- Descrizione:Roller for cooling boxes