Orders LinkContactUsLinkComponent
Thermosafe Cool Boxes | Avantor
Thermosafe Cool Boxes
Roller for cooling boxes, -
Katalognr. 216-7002
Anbieter:   HONIGBERG
Thermosafe Cool Boxes
Katalognr. 216-7002
Anbieter:  HONIGBERG
Thermosafe Cool Boxes
Thermosafe Cool Boxes
Roller for cooling boxes, -
Preis
SFr. 1’490.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PE, grey.
  • Insulated with 5,75 cm thick plastic foam
  • Hinged lid with robust hinges and metal catch, one sturdy carrying handle with the 35 litre model and two with the 75 litre model
  • Aluminium roller base, with 4 moving rollers, available separately
Large capacity cool boxes for the long-term temperature control of temperature-sensitive materials and samples. Cooled with cooling pouches 216-7010 to 216-7013 or dry ice.
Technische Daten
  • Capacità:
    -
  • Descrizione:
    Roller for cooling boxes