Dropping bottles, Lucas
15 ml
Catalog # 215-9102
Supplier: MUELLER KREMPEL
Jump to:
Product Details & Documents
Amber glass, without closure.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.
Specifications
- Capacità:15 ml
- Seq:04