Sample bottles with screw cap
4,50 ml, 14×50 mm, Yes
Codice catalogo 215-8343
Nome del fornitore: MUELLER KREMPEL
Vai a
Product Details & Documents
Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.
- Thin-walled, cylindrical
- Tightly sealing screw cap
- Designed for use in applications that do not require resistance to temperature
Specifiche
- Tappo incluso:Yes
- Ø×A:14×50 mm
- Capacità:4,50 ml
- Cat. No.:215-8343