Sample bottles with screw cap
1,50 ml, 13×27 mm, Yes
Catalog # 215-8342
Supplier: MUELLER KREMPEL
Jump to:
Product Details & Documents
Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.
- Thin-walled, cylindrical
- Tightly sealing screw cap
- Designed for use in applications that do not require resistance to temperature
Specifications
- Tappo incluso:Yes
- Ø×A:13×27 mm
- Capacità:1,50 ml