Orders LinkContactUsLinkComponent
Canonical URL fetch failed:ERROR
Replacement caps
SVL 25
Catalog # 215-2046
Supplier:   QVF LABORTECHNIK
Replacement caps
Catalog # 215-2046
Supplier:  QVF LABORTECHNIK
Replacement caps
SVL 25
Price
SFr. 9.50
Log in for availability
1 * 1 ST
Product Details & Documents
Pyrex®, phenoplast, black.
  • Excellent chemical resistance, excellent temperature resistance
  • Screw caps with PTFE-coated seal
  • Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.
Specifications
  • Filettatura:
    SVL 25