Replacement caps
SVL 18
Codice catalogo 215-2043
Nome del fornitore: QVF LABORTECHNIK
Vai a
Product Details & Documents
Pyrex®, phenoplast, black.
- Excellent chemical resistance, excellent temperature resistance
- Screw caps with PTFE-coated seal
- Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.
Specifiche
- Filettatura:SVL 18