Measuring cylinders, conical
1000 ml, 140 mm
Catalog # 213-8021
Supplier: CONTACTO
Jump to:
Product Details & Documents
Stainless steel, with handle.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Specifications
- Capacità:1000 ml
- Altezza:140 mm
- Seq:02