Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
11 mm, 24 (2×12), 125×42×35 mm
Codice catalogo 212-6310
Nome del fornitore:   SCHAEFER GUENTHER KUNSTSTOF
Racks for test tubes
Codice catalogo 212-6310
Nome del fornitore:  SCHAEFER GUENTHER KUNSTSTOF
Racks for test tubes
Racks for test tubes
11 mm, 24 (2×12), 125×42×35 mm
Price
166.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Rack for test tubes, made from clear acrylic, constructed in two tiers.
  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
Specifiche
  • Ø foro:
    11 mm
  • N. di fori:
    24 (2×12)
  • L×P×A:
    125×42×35 mm