Rack per PCR
Rack per PCR con 96 pozzetti, colori assortiti
Codice catalogo 211-3505
Nome del fornitore: HEATHROW
Vai a
Product Details & Documents
High resistance PP.
- Tube wells are easily identified by imprinted numbers and letters
- Removable, hinged lid
Designed to accommodate 96 individual 0,2 ml tubes or 12 or 8 strips of 8 or 12 tubes.
Colour: Natural, blue, green, yellow or assorted colours (one each of blue, green, pink, yellow and natural).
Specifiche
- Colore:colori assortiti
- Descrizione:Rack per PCR con 96 pozzetti