Orders LinkContactUsLinkComponent
Rack per PCR
Rack per PCR con 96 pozzetti, naturale
  211-3504
 :   HEATHROW
Rack per PCR
  211-3504
 :  HEATHROW
Rack per PCR
Rack per PCR
Rack per PCR con 96 pozzetti, naturale
SFr. 10.00
1 * 1 ST
High resistance PP.
  • Tube wells are easily identified by imprinted numbers and letters
  • Removable, hinged lid
Designed to accommodate 96 individual 0,2 ml tubes or 12 or 8 strips of 8 or 12 tubes.
Colour: Natural, blue, green, yellow or assorted colours (one each of blue, green, pink, yellow and natural).
  • Colore:
    naturale
  • Descrizione:
    Rack per PCR con 96 pozzetti