Spazzole da laboratorio
Set 1, Martora rossa, —
149-2127
Marketsource
This brush range covers a wide variety of applications thanks to the number of styles on offer.
- Paintbrush made of pale ox hair
- Red marten brush — pure marten hair with black painted wooden handle
- Goat-hair brush — oval, on wooden handle
- Wide bear-hair brush, flat handle
Set 1: Contains 1 red marten brush in each size, sizes 1 to 5.
- Materiale setola:Martora rossa
- Testa della spazzola Ø:—
- Descrizione:Set 1
- Seq:15