Orders LinkContactUsLinkComponent
Labels, hazardous substance
Labels, hazardous substance
Hazard labels, 26×26 mm
Catalog # 116-1571
Supplier:   LAB IN
Labels, hazardous substance
Catalog # 116-1571
Supplier:  LAB IN
Labels, hazardous substance
Hazard labels, 26×26 mm
Price
SFr. 6.60
Log in for availability
1 * 10 ST
Product Details & Documents
Hazardous substance labels from self- adhesive foil for containers.
  • Available in three dimensions
Acc. GefStoffV.
Specifications
  • Descrizione:
    Hazard labels
  • Dimensioni:
    26×26 mm
  • Seq:
    31