Agarose I, Pierce™
Agarose I
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Agarose I (molecular biology grade) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparation of electrophoresis gels.
- Standard melting temperature agarose for electrophoresis
- Separation range: 100 bp to >30 kb
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Descrizione:Agarose I
- Seq:01