Orders LinkContactUsLinkComponent
Spider evaporators | Avantor
Spider evaporators
Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint
Codice catalogo BUCH011577
Spider evaporators
Codice catalogo BUCH011577
Nome del fornitore:  
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint
Discontinued
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifiche
  • Descrizione:
    Spider with cylindric flasks, SJ24/40, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
  • Tipo:
    For simultaneous distillation from 12×20 ml cylindrical flasks with 24/40 NS Joint