Evaporating flasks
100 ml, 24/29 NS, uncoated
Codice catalogo BUCH000473
Nome del fornitore: BUCHI
Vai a
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifiche
- Cono femmina:24/29 NS
- Tipo:uncoated
- Capacità:100 ml
- Seq:04