Cold traps with/without condensation drain
200 ml, without condensate drain
Catalog # 181-9982
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump.
DURAN®, chiaro.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Descrizione:200 ml, without condensate drain
- Seq:01