Distillatori
400 mm, 29/32 NS, 14/23 NS
Codice catalogo 201-3132
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Bridge includes fused Liebig condenser and curved vacuum adapter. NS 14/23 socket for distillation thermometer with 55 mm fitted length. Water and vacuum connection via hose connectors with 8 mm external diameter or water connection with thread GL 14 and vacuum receiver adapter with thread GL 18.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Schott tubing
Scheda dati
Specifiche
- Dimensione cono:29/32 NS
- Lunghezza mantello:400 mm
- Cono femmina:14/23 NS



