Adapters, expansion, DURAN®
Expansion, 24/29 NS, 29/32 NS
Codice catalogo SCOT286548201
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3,3 clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Specifiche
- Dimensione cono:24/29 NS
- Cono femmina:29/32 NS
- Tipo:Expansion
- Seq:07