Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
4 mm, 18,8 NS
Codice catalogo SCOT286459209
Nome del fornitore:   DWK Life Sciences
Stopcocks, single way, DURAN®
Codice catalogo SCOT286459209
Nome del fornitore:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
4 mm, 18,8 NS
Price
1 160.00 SFr.
Log in per disponibilità
1 * 25 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with hollow glass key and glass handle.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Specifiche
  • Ø foro:
    4 mm
  • Seq:
    04
  • Rubinetto:
    18,8 NS