Stopcocks, single way, DURAN®
4 mm, 18,8 NS
Codice catalogo SCOT286459209
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with hollow glass key and glass handle.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifiche
- Ø foro:4 mm
- Seq:04
- Rubinetto:18,8 NS