Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
10 mm, 29,2 NS
Codice catalogo SCOT286441607
Nome del fornitore:   DWK Life Sciences
Stopcocks, single way, DURAN®
Codice catalogo SCOT286441607
Nome del fornitore:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
10 mm, 29,2 NS
Price
359.00 SFr.
Log in per disponibilità
1 * 5 ST
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.
Specifiche
  • Ø foro:
    10 mm
  • Rubinetto:
    29,2 NS