Stopcocks, single way, DURAN®
8 mm, 24 NS
Codice catalogo SCOT286441401
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
- Very good chemical resistance
- Very good temperature resistance
- For glass manipulators
Ungrounded, without key.
Specifiche
- Ø foro:8 mm
- Rubinetto:24 NS