Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
6 mm, 21,5 NS
Codice catalogo SCOT286441007
Nome del fornitore:   DWK Life Sciences
Stopcocks, single way, DURAN®
Codice catalogo SCOT286441007
Nome del fornitore:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
6 mm, 21,5 NS
Price
SFr. 218.00
Log in per disponibilità
1 * 5 ST
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.
Specifiche
  • Ø foro:
    6 mm
  • Rubinetto:
    21,5 NS