Stopcocks, single way, DURAN®
2 mm, 12,5 NS
Codice catalogo SCOT286439001
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
- Very good chemical resistance
- Very good temperature resistance
- For glass manipulators
Ungrounded, without key.
Specifiche
- Ø foro:2 mm
- Rubinetto:12,5 NS