Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
2 mm, 12,5 NS
Codice catalogo SCOT286439001
Nome del fornitore:   DWK Life Sciences
Stopcocks, single way, DURAN®
Codice catalogo SCOT286439001
Nome del fornitore:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
2 mm, 12,5 NS
Price
SFr. 966.00
Log in per disponibilità
1 * 50 ST
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.
Specifiche
  • Ø foro:
    2 mm
  • Rubinetto:
    12,5 NS