Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
18,8 NS
  SCOT286433804
Stopcocks for separating funnels, DURAN®
  SCOT286433804
Stopcocks for separating funnels, DURAN®
18,8 NS
1 290.00 SFr.
1 * 25 ST
DURAN®, borosilicate glass, with polished short funnel, without key.
  • Very good chemical resistance
  • High temperature resistance
For glass manipulators.
  • Rubinetto:
    18,8 NS