Stopcocks for separating funnels, DURAN®
18,8 NS
Catalog # SCOT286426809
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
- Very good chemical resistance
- Very good temperature resistance
For glass manipulators.
Specifications
- Rubinetto:18,8 NS