Stopcocks, single-way, DURAN®
2 mm, 12,5 NS
Codice catalogo 201-0383
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifiche
- Ø foro:2 mm
- Seq:18
- Rubinetto:12,5 NS