Stopcocks, single-way, DURAN®
2,5 mm, 14,5 NS
Catalog # SCOT286412009
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifications
- Ø foro:2,5 mm
- Seq:16
- Rubinetto:14,5 NS