Orders LinkContactUsLinkComponent
Stopcocks for burettes, angled, DURAN®
Stopcocks for burettes, angled, DURAN®
with PTFE key, 2,5 mm, 12,5 NS
Catalog # SCOT286409607
Supplier:   DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Catalog # SCOT286409607
Supplier:  DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Stopcocks for burettes, angled, DURAN®
with PTFE key, 2,5 mm, 12,5 NS
Price
SFr. 1’780.00
Log in for availability
1 * 50 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, with glass key.
  • Very good chemical resistance
  • Very good temperature resistance
  • Screwthread retaining device
Specifications
  • Ø foro:
    2,5 mm
  • Descrizione:
    with PTFE key
  • Rubinetto:
    12,5 NS