Stopcocks for burettes, angled, DURAN®
with PTFE key, 2,5 mm, 12,5 NS
Catalog # SCOT286409607
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, with glass key.
- Very good chemical resistance
- Very good temperature resistance
- Screwthread retaining device
Specifications
- Ø foro:2,5 mm
- Descrizione:with PTFE key
- Rubinetto:12,5 NS