Stopcocks for burettes, angled, DURAN®
with SBW glass key, 2,5 mm, 12,5 NS
Codice catalogo SCOT286409204
Nome del fornitore: DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Codice catalogo SCOT286409204
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, with glass key.
- Very good chemical resistance
- Very good temperature resistance
- Screwthread retaining device
Specifiche
- Ø foro:2,5 mm
- Descrizione:with SBW glass key
- Rubinetto:12,5 NS