Screwthread tubes, attachable, DURAN®
GL 45, 40 mm, 2,3 mm
Codice catalogo SCOT286063403
Nome del fornitore: DWK Life Sciences
Screwthread tubes, attachable, DURAN®
Codice catalogo SCOT286063403
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Caution: These tubes are open at the bottom.
Specifiche
- Filettatura:GL 45
- Ø esterno tubo:40 mm
- Spessore parete:2,3 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



