Orders LinkContactUsLinkComponent
DURAN® Blanks for Evaporating Flasks
500 ml
Catalog # SCOT210284407
Supplier:   DWK Life Sciences
DURAN® Blanks for Evaporating Flasks
Catalog # SCOT210284407
Supplier:  DWK Life Sciences
DURAN® Blanks for Evaporating Flasks
DURAN® Blanks for Evaporating Flasks
500 ml
Price
SFr. 287.00
Log in for availability
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.
Specifications
  • Capacità:
    500 ml
  • Seq:
    04