DURAN® Blanks for Evaporating Flasks
500 ml
Catalog # SCOT210284407
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.
Specifications
- Capacità:500 ml
- Seq:04