DURAN® Blanks for Evaporating Flasks
100 ml
Codice catalogo SCOT210282406
Nome del fornitore: DWK Life Sciences
DURAN® Blanks for Evaporating Flasks
Codice catalogo SCOT210282406
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.
Specifiche
- Capacità:100 ml
- Seq:01