Bottiglie per burette automatiche
2000 ml, Reservoir bottle, clear glass
Codice catalogo 612-3219
Nome del fornitore: Brand
Vai a
Product Details & Documents
Borosilicate glass 3.3.
- Very good temperature resistance
- Very good chemical resistance
- Amber coloured for light sensitive substances
- Socket size NS 29732
Reservoir bottle for automatic burette available in clear glass or amber glass.
Specifiche
- Capacità:2000 ml
- Descrizione:Reservoir bottle, clear glass