Accessories for Spectroquant® NOVA
Software for Windows (in English and German) for the data transfer, NOVA photometers Spectroquant®
Nome del fornitore: Merck
Vai a
Product Details & Documents
To document the analysis values, either a standard computer or a printer can be connected.
- The Multi-Agate II software can control data acquisition or simply record recorded analysis data
- The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.
Documents
msds_100787 msds_109749 msds_109769 msds_114667 msds_114723
Scheda di sicurezza
More
Specifiche
- Descrizione:Software for Windows (in English and German) for the data transfer
- Per:NOVA photometers Spectroquant®