Rack per microprovette, quattro vie, PP, FLIPPER™
rosa fluorescente, 0,5/1,5/15/50 ml
Catalog # 700-2851
Supplier: Thermo Fisher Scientific
Rack per microprovette, quattro vie, PP, FLIPPER™
Catalog # 700-2851
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Made of PP.
- Excellent chemical resistance
- Excellent temperature resistance
- Sturdy
These racks are for microcentrifuge tubes with sides having frosted writing surface for smear-resistant labelling.
Specifications
- Per provette:0,5/1,5/15/50 ml
- Colore:rosa fluorescente
- Seq:03
- Cat. No.:700-2851
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)