Burettes, lateral stopcock, class AS, Schellbach
50 ml, 0,10 ml, 0,05 ± ml
Codice catalogo 661-4122
Nome del fornitore: witeg Labortechnik
Burettes, lateral stopcock, class AS, Schellbach
Codice catalogo 661-4122
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Burettes, class AS, Schellbach blue line, conformity certified and made of clear borosilicate glass.
- Main point ring graduation
- Lateral stopcock, with PTFE-needle valve
- Overtwist device
Specifiche
- Capacità:50 ml
- Divisione:0,10 ml
- Tolleranza:0,05 ± ml



