Pipette tips, ratiolab®
Type E, 100 - 1200 µl, -, Blu, commonly used pipettes, 1000/sacchetto
Codice catalogo 612-2501
Nome del fornitore: RATIOLAB
Vai a
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifiche
- Confezione:1000/sacchetto
- Sterili:-
- Volume:100 - 1200 µl
- Seq:29
- Stile punta:Type E
- Colore puntale:Blu
- Per uso con:commonly used pipettes