Pipette Pasteur
Graduated, 1,0 ml, 40 µl, 138 mm, -, Bulk
Catalog # 612-1740
Supplier: RATIOLAB
Jump to:
Product Details & Documents
In PE, monouso
- Soft, elastic bulb
- Unbreakable, reduced risk of injury compared to glass
- Inert to biological fluids and most acids
- Graduated pipettes with clearly visible embossed graduations
- Sterile pipettes are gas-sterilised and individually packed
- Suitable for food contact
Specifications
- Confezione:Bulk
- Sterili:-
- Capacità:1,0 ml
- Descrizione:Graduated
- Lunghezza:138 mm
- Seq:06
- Volume gocce:40 µl