Pipette tips, ratiolab®
Oxford, 1 - 200 µl, -, Trasparente, Pipette Oxford, 1000/sacchetto
Catalog # 612-1480
Supplier: RATIOLAB
Jump to:
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifications
- Confezione:1000/sacchetto
- Sterili:-
- Volume:1 - 200 µl
- Seq:42
- Stile punta:Oxford
- Colore puntale:Trasparente
- Per uso con:Pipette Oxford