Tappi per colonna
Column end plug, black
Codice catalogo 554-1146
Nome del fornitore: VICI JOUR RESEARCH
Vai a
Product Details & Documents
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
- Available with a 5/16" wrench flat
- Easy removal
Specifiche
- Descrizione:Column end plug, black
- Seq:02