Screw caps with liners, WHEATON®
Tappi a vite, finitura filettata GPI 24 - 400, White PP, solid top, Foamed PE
Codice catalogo 548-2043
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
These PP screw caps have multi-purpose liners, which are made of three-ply co-extruded foamed low density PE core between two solid layers of low density PE.
- For use with neutral pH to mild acid and alkali solutions
Specifiche
- Setto:Foamed PE
- Tipo:White PP, solid top
- Descrizione:Tappi a vite, finitura filettata GPI 24 - 400
- Seq:04