Jars, with lid
100 mm, 50 mm
Codice catalogo 525-1362
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Specifiche
- Ø est:100 mm
- Altezza:50 mm
Jars, low form, made of glass, with notch and overlapping cover.