Test tube baskets
229×229×229 mm
Codice catalogo 489-2212
Nome del fornitore: Bel-Art Products, a Part of SP
Vai a
Product Details & Documents
In PP.
- Very good chemical resistance
- Very good temperature resistance
- Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White
Specifiche
- L×P×A:229×229×229 mm