Test tube baskets
152×152×152 mm
Catalog # 489-2211
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
In PP.
- Very good chemical resistance
- Very good temperature resistance
- Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White
Specifications
- L×P×A:152×152×152 mm